Sodium-5-(2-aminoethylamino)-1-naphthalenesulfonate hydrate ≥95%

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Sodium N-(2-Aminoethyl)-5-naphthylamine-1-sulphonate, 5-(2-Aminoethylamino)-1-naphthalenesulphonic acid Sodium Salt, N-(2-Aminoethyl)-5-naphthylamine-1-sulphonic acid Sodium Salt, 1,5-EDANS sodium salt, EDANS, sodium salt

616
76480-996EA 142.55 USD
76480-996 76480-998
Sodium-5-(2-aminoethylamino)-1-naphthalenesulfonate hydrate ≥95%
Sodium-5-(2-aminoethylamino)-1-naphthalenesulfonate hydrate

EDANS sodium salt is much more water-soluble than EDANS acid.

Formula: C₁₂H₁₄N₂O₃S
MW: 288.30 g/mol
MDL Number: MFCD00051474
CAS Number: 100900-07-0
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e07330012b19671c95d43eb3b22c178a2a75f44&vid=c3d56bdb9772fa36efe073bdb8e253763bfa7e21&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR