VWR® P2-Series Balances
Supplier: VWR International
76175-276 76175-278 76175-280 76175-398 76175-400 76175-402 76175-404 76480-252 76480-254 76480-256 76480-258 76480-260
76175-276EA 813.55 USD
76175-276 76175-278 76175-280 76175-398 76175-400 76175-402 76175-404 76480-252 76480-254 76480-256 76480-258 76480-260
VWR® P2-Series Balances
Balances and Scales
VWR® P2-Series Balances combine portability and accuracy in a compact package.
  • Worry-free five-year warranty
  • Reliable results
  • External and internal calibration options
  • Multiple weigh modes
  • Backlit, LCD screen
  • Internal, rechargeable battery
Operate in the field or move from lab-to-lab. The VWR® P2-Series come complete with a power adapter and an internal rechargeable battery. Automatic shut off mode conserves battery life and the low battery indicator ensures sample weighing is not interrupted.
These economical balances offer a variety of standardMore Product Information