Oxygen Pressure Regulator, PerkinElmer
Supplier: PerkinElmer
02400084
101687-698EA 1162.66 USD
101687-698 76474-984
Oxygen Pressure Regulator, PerkinElmer
Valves Pressure-Relief Valves
Gas Pressure Regulator Features: Barstock Body Construction Barstock bodies have a lower internal volume than forged bodies, allowing for more complete purging of atmospheric contaminants. Stainless Steel Diaphragms Unlike elastomeric diaphragms, stainless steel diaphragms don't absorb atmospheric contaminants or allow them to diffuse into the gas stream. Metal-to-metal Seals Metal-to-metal seals provide reliable protection from inboard leakage of atmospheric contaminants.