Static Dissipative Wipes, Transforming Technologies
Supplier: Transforming Technologies
WP6200
76474-830EA 99.13 USD
76474-830
Static Dissipative Wipes, Transforming Technologies
Wipes Cleanroom Wipes Dry Cleanroom Wipes
ESD and cleanroom safe, these static dissipative wipes are perfect for all electronic manufacturing and optical cleaning applications.
  • 100% knitted polyester
  • No carbon or metal fibers
  • Good abrasion and solvent resistance
  • Lint-free
ESD safe wipers are made from clean, durable synthetic fibers and their substrate is highly absorbent and will not turbocharge over 100 V. The wipes are perfect for optics and LCD screen cleaning because they contain no carbon or metal fibers which can scratch vulnerable surfaces.