Ptec™ Self-Cleaning Bench Top Ionizer, Transforming Technologies
Supplier: Transforming Technologies
IN5500
76474-666EA 952.34 USD
76474-666
Ptec™ Self-Cleaning Bench Top Ionizer, Transforming Technologies
Ionizers
The Ptec™ IN5500 AC benchtop air ionizer is a small, efficient static elimination device with variable fan speed for protecting electronic components and general static protection.
  • Remove static charges quickly and efficiently
  • Self-cleaning system simplifies service and maintenance
  • Perfect for single station table top, bench, or desk use
  • Balance: ±10 V
  • Small footprint
The unit can self-clean, which eliminates maintenance costs and equipment downtime and ensures the ionizer provides optimal static eliminating performance. Using AC corona technology toMore Product Information