Stuart® Magnetic Hotplate Stirrers, Digital, Undergrad Series, Cole-Parmer
Supplier: Antyila Scientific
04807-63 04807-65
76474-330EA 589.25 USD
76474-330 76474-332
Stuart® Magnetic Hotplate Stirrers, Digital, Undergrad Series, Cole-Parmer
Stirrers Magnetic Hotplate Stirrers
Get true temperature setting and accurate stirring control.
  • Hot warning light remains on even when unplugged
  • BioCote® glaze for antimicrobial protection
  • Tough aluminum casing for longevity
  • Designed to prevent accidental spillage on control panel
  • Compact, stackable design takes up less benchtop and storage space
  • IP 32 rated for water and dust resistance
Designed with microprocessor-controlled dual thermocouples, these magnetic hotplate stirrers offer accurate temperature control and protection from over-heating. Actual surface temperature is displayedMore Product Information