Sterilization Wraps for Steam, ETO or Gamma, Blue, Heavy Duty, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BHDSHEET0909 BHDSHEET1515 BHDSHEET1818 BHDSHEET2020 BHDSHEET2424 BHDSHEET3030 BHDSHEET3636 BHDSHEET4040 BHDSHEET4545 BHDSHEET5454 BHDSHEET5472
76471-676CS 446.62 USD
76471-676 76471-680 76471-682 76471-684 76471-686 76471-688 76471-690 76471-692 76471-694 76471-696 76471-698
Sterilization Wraps for Steam, ETO or Gamma, Blue, Heavy Duty, Keystone Cleanroom Products
Sterilization Wraps
When secured with autoclave tape or elasticized bands, these blue BHD wrappers provide an effective microbial barrier from external sources of contamination.
  • Designed for steam sterilization; but also compatible with ETO or gamma irradiation
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Sufficient microbial barrier
  • Superior bacterial barrier properties compared to linen
  • Sterilization tapes adhere firmly and remove easily
  • Ideal choice for
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable