Spatulas, Double End Mixing, Excelta Corp®
Supplier: Excelta
344
63041-976EA 12.57 USD
63041-976 63041-980 76471-554
Spatulas, Double End Mixing, Excelta Corp®
Spatulas
These stainless steel spatulas are used for applying or scraping potting compound or wax.
  • Double ended for flexibility
  • Cleanroom safe, stainless steel
These large spatulas are primarily used for applying or scraping potting compound or wax. They can also be used for mixing epoxy.The 342 and 343 are made of stainless steel, 7" long.
Order Now
SPECIFICATIONS
Environmentally Preferable