VWR® SMS Sleeves, Made in USA
Supplier: VWR International
76470-786
76470-786CS 461.94 USD
76470-786
VWR® SMS Sleeves, Made in USA
Disposable Garments
VWR® Made in USA SMS Sleeves are an ideal option for single-use needs.
  • 3-layer SMS construction
  • Serged seams
  • Produced with latex-free elastic
  • Made in USA
A key part of the VWR® Made in USA PPE line, these SMS sleeves are 20" length to provide a comfortable fit while meeting requirements of many controlled environments.
Envase: 100 per case. Packed in poly-lined bag within brown corrugated box.