VWR® Sterile Cleanroom Face Masks
Supplier: VWR International
76470-110 76470-112
76470-110CS 381.04 USD
76470-110 76470-112
VWR® Sterile Cleanroom Face Masks
Masks Hygiene Masks
Sterile cleanroom face masks are designed for critical applications in cleanroom, biotech, pharmaceutical, and manufacturing environments. Offering 99.7% BFE and 99.79% PFE with a Delta P of 4.0 (mm H₂O/cm²), these masks meet ASTM standards for superior filtration and breathability.
  • Ultrasonically welded
  • Three-layer construction: 30 gsm polypropylene, 27 gsm filtration media, 25 gsm polypropylene
  • One size fits all
  • Gamma sterilized
Made with a three layer construction (polypropylene and filtration media), masks feature a metal nosepiece for a secure
More Product Information