sparQ UDI Adapters (1-96), Quantabio
Supplier: Quantabio
95211-096
76468-936EA 946.29 USD
76468-936
sparQ UDI Adapters (1-96), Quantabio
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
sparQ UDI Adapters (1-96) are unique dual-indexed adapters designed to be used on illumina sequencing platforms. It is compatible with both DNA and RNA libraries. The use of UDI’s minimizes index hopping events while also enabling greater multiplexing on flow cells.
  • Flexible pooling: Multiplex up to 96 samples per sequencing run
  • Improved performance: Prevents index hopping and enhances demultiplexing accuracy
  • Optimized: For maximum ligation efficiency with sparQ library prep kits
  • Multiple applications: Including whole genome (with amplification or PCR-free), target enrichment, whole transcriptome sequencing and many more