Anti-ACACB Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
A03668-2
76465-220EA 407.19 USD
76465-220
Anti-ACACB Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Acetyl Coenzyme A Carboxylase/ACACB detection. Tested with WB, IHC-P, FCM in Human,Mouse,Rat.

Type: Primary
Antigen: ACACB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,Mouse,Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR