SatPax® MicroSeal®-VP Pre-Wetted Sealed Edge Cleanroom Laundered Wipes 100% Polyester, Berkshire
Supplier: Berkshire
SPXVP00112
76461-872CS 351.37 USD
76461-872
SatPax® MicroSeal®-VP Pre-Wetted Sealed Edge Cleanroom Laundered Wipes 100% Polyester, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Lightweight, high absorbency material design reduces actual cost in use compared to heavier weight products.
  • Critically low particles, fibers, ions, and extractables
  • Pre-wetted with 70% USP IPA/30% DI water
  • High abrasion resistance
  • High absorbency
  • Increases cleaning protocol consistency
  • Ultrasonically sealed edge for reduced fiber contamination
  • 100% continuous filament polyester knit
SatPax® MicroSeal®-VP is an ultrasonically sealed edge cleanroom laundered wiper recommended for ISO Class 3 and above environments. It is composed of 100% continuousMore Product Information