High Temperature 8 to 4 Gauge Wire Marking Sleeves, 3" Core, Brady
Supplier: Brady Worldwide
2HT-375-2-WT-S
76461-298RL 1079.86 USD
76461-298
High Temperature 8 to 4 Gauge Wire Marking Sleeves, 3" Core, Brady
Labels
PermaSleeve® Heat-Shrink Polyvinylidene Fluoride (B-345) wire marker sleeves are specifically designed for high temperature and low vacuum outgassing applications.
  • Flame-retardant, heat-shrinkable
  • Double-side printable
PermaSleeve® 2:1 heat shrinkable sleeves are fade-resistant and flame-retardant, and meet the requirements of NASA Vacuum Outgassing specification SP-R-0022A. Sleeves can be used in applications that require greater resistance to harsh chemicals such as fuels, lubricants, and high power cleaning solvents. Operating temperature range of ‒More Product Information