High Temperature Flame Retardant Wire Wraps, 3" Core, Brady
Supplier: Brady Worldwide
THT-106-472
76461-010RL 867.04 USD
76461-010
High Temperature Flame Retardant Wire Wraps, 3" Core, Brady
Labels
Flame Retardant Wire Wraps (B-472) are ideal for cable and wire bundle applications and label applications where self-extinguishing properties are required.
  • Flame-retardant
Polyimide material specifically designed to self-extinguish and prevent flames from propagating. Matte finish material with permanent acrylic adhesive can be thermal transfer printed or dot matrix printed. Service temperature of –40 to +212 °F (–40 to +100 °C).
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® SeriesMore Product Information