PrintMate™ AS Cassette Printer Total Care Packages, Epredia™
Supplier: Epredia
A83800001B A83800002B A83800003B A83800004B A83800005B A83800006B
76459-074EA 66652.54 USD
76459-074 76459-076 76459-078 76459-080 76459-082 76459-084
PrintMate™ AS Cassette Printer Total Care Packages, Epredia™
Printers Cassette and Slide Printers
The PrintMate™ AS Total Care Package offers workflow protection by offering a complete solution, combining the PrintMate™ AS on-demand cassette printer with cassettes and total care service support.
  • The entire range of cassettes has been redesigned for better performance – both in printing and in laboratory processes
  • The all-new surface finish and reinforced print faces are built to optimize print quality when used with PrintMate AS cassette printers
  • All-new PrintMate™ AS-compatible tubes are precisely engineered to reduce jamming and prevent downtime
More Product Information