VWR® Sterile Class 100 HS Nitrile Gloves
Supplier: VWR International
76458-114 76458-116 76458-118 76458-120 CETSTERILE5 76458-124 76458-126
76458-114CS 368.38 USD
76458-114 76458-116 76458-118 76458-120 76458-122 76458-124 76458-126
VWR® Sterile Class 100 HS Nitrile Gloves
Gloves
The VWR® sterile Class 100 HS nitrile gloves are intended for use in cleanroom production and research environments. These hand-specific gloves are pair packed for easy donning.
  • Gamma irradiated to SAL 10⁻⁶
  • Made from premium compounded Nitrile
  • Single use only
  • Suitable for use in ISO Class 5 (FED-STD-209E Class 100) or higher cleanrooms
Manufactured from a synthetic copolymer of acrylonitrile, butadiene, and methylacrylic acid (components that comply with FDA CFR 21). High tensile strength and strong permeation resistance against a wide variety ofMore Product Information