Tyvek® 1073B Self-Sealing / PE Steam Sterilization Chevron Peel Pouches with Steam Indicator, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BBMGTVKSS0206 BBMGTVKSS02095 BBMGTVKSS0306 BBMGTVKSS0308 BBMGTVKSS03508 BBMGTVKSS0415 BBMGTVKSS0422 BBMGTVKSS0510 BBMGTVKSS0608 BBMGTVKSS0612 BBMGTVKSS0628 BBMGTVKSS0632 BBMGTVKSS0635 BBMGTVKSS0713 BBMGTVKSS0810 BBMGTVKSS0815 BBMGTVKSS0818 BBMGTVKSS0834 BBMGTVKSS1012 BBMGTVKSS1020 BBMGTVKSS1214 BBMGTVKSS1217 BBMGTVKSS1424 BBMGTVKSS1520 BBMGTVKSS1617 BBMGTVKSS1626 BBMGTVKSS1632 BBMGTVKSS1635 BBMGTVKSS1635X BBMGTVKSS1822 BBMGTVKSS2426 BBMGTVKSS2430 BBMGTVKSS2435
76450-870CS 1314.1 USD
76450-870 76450-872 76450-874 76450-876 76450-878 76450-880 76450-882 76450-884 76450-886 76450-888 76450-890 76450-892 76450-894 76450-896 76450-898 76450-900 76450-902 76450-904 76450-906 76450-908 76450-910 76450-912 76450-914 76450-916 76450-918 76450-920 76450-922 76450-924 76450-926 76450-928 76450-930 76450-932 76450-934
Tyvek® 1073B Self-Sealing / PE Steam Sterilization Chevron Peel Pouches with Steam Indicator, Keystone Cleanroom Products
Bags Sterilization Pouches
Improve the prep time, performance, microbial barrier and cleanliness of your autoclave peel pouches.
  • Manufactured and packaged in a controlled cleanroom environment
  • Easy, fast and reliable self-sealing closure system
  • Autoclavable at 121 to 125 °C
  • Peels open cleanly, chevron peel, hydrophobic
  • Tear and puncture resistant, lint-free
  • Integrated steam sterilization indicator
A significant upgrade over paper based steam sterilization pouches. These Tyvek® peel pouches will not rip or puncture like paper based autoclave pouches routinely do. Save valuableMore Product Information