Saliva Collection Kit for Covid-19 Sample Collection, IBI Scientific
Supplier: IBI Scientific
SK-550
76449-908CS 2917.82 USD
76449-908
Saliva Collection Kit for Covid-19 Sample Collection, IBI Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Sputum Sample Collection
For COVID-19 saliva sample collection.
  • No storage buffers or lysis reagents included (dry kit)
  • Tubes are barcoded for individual sample identification
  • Tubes are compatible with robotic filling systems
All kits, tubes and over-packs are labeled for Lot Control for traceability.
Información de suministro: Individual kit includes instructions, collection tube with cap, collection funnel that fits tube.