VWR® Calibration Weights
Supplier: VWR International
VWR-E2-50G VWR-E2-100G VWR-E2-200G VWR-F1-100G VWR-F1-200G VWR-F1-500G VWR-F1-1KG VWR-F1-2KG VWR-F1-5KG VWR-F2-500G VWR-F2-1KG VWR-F2-2KG VWR-F2-5KG VWR-F2-10KG VWR-F2-20KG VWR-E2-50G/C VWR-E2-100G/C VWR-E2-200G/C VWR-F1-100G/C VWR-F1-200G/C VWR-F1-500G/C VWR-F1-1KG/C VWR-F1-2KG/C VWR-F1-5KG/C VWR-F2-500G/C VWR-F2-1KG/C VWR-F2-2KG/C VWR-F2-5KG/C VWR-F2-10KG/C VWR-F2-20KG/C
76404-666EA 117.03 USD
76404-666 76404-668 76404-670 76404-672 76404-674 76404-676 76404-678 76404-680 76404-682 76404-684 76404-686 76404-688 76404-690 76404-692 76404-694 76446-828 76446-830 76446-832 76446-834 76446-836 76446-838 76446-840 76446-842 76446-844 76446-846 76446-848 76446-850 76446-852 76446-854 76446-856
VWR® Calibration Weights
Weights Calibration Weights
VWR® Calibration Weights are crafted from high grade stainless steel and feature a plastic storage case.
  • Option to be supplied with a Statement of Accuracy or an ISO 17025:2017 Certificate of Calibration
  • Cylindrical with knob
  • OIML Class E2, F1, or F2 options available
  • High polish finish
  • Low magnetic permeability
The ISO 17025:2017 Certificate of Calibration provides full traceability of the calibration weight. This certificate provides the date of manufacture, serial number, nominal mass, conventional mass, and uncertainty of measurement.