EVA PAPR with 20LF2HE Loose-fitting Facepiece, Complete System, Bullard
Supplier: Bullard
EVAPAN4
76443-020EA 1788.95 USD
76443-020
EVA PAPR with 20LF2HE Loose-fitting Facepiece, Complete System, Bullard
Respirators Respiratory Systems
World Class PAPR that is ready to use directly out of the box.
  • All-in-one system ready for use, including one HEPA filter
  • Filters exhalated air to keep patients safe
  • Assigned protection factor (APF) 2.5X greater than an N95 mask
  • Improves breathing and overall comfort compared to an N95 mask
  • Loose-fitting facepiece designed for use with a stethoscope
  • No fit test
  • HEPA filter for use against communicable disease
EVA Powered Air-Purifying Respirator (PAPR) is equipped with the latest technology for providing wearers with constant airflow in a compact,More Product Information