Avantor® Hichrom HI-DEX DET Gamma, GC Columns
Supplier: VWR International
HI69-25-015-15 HI69-25-015-25 HI69-25-015-30 HI69-25-015-50 HI69-25-015-60 HI69-25-025-15 HI69-25-025-25 HI69-25-025-30 HI69-25-025-50 HI81-10-010-10 HI81-10-010-5
76429-954EA 1550.26 USD
76429-954 76429-956 76429-958 76429-960 76429-962 76429-964 76429-966 76429-968 76429-970 76429-972 76430-074 76430-076
Avantor® Hichrom HI-DEX DET Gamma, GC Columns
Chromatography Columns
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Order Now
SPECIFICATIONS
Phase name HI-DEX DET Gamma
Environmentally Preferable
Phase composition Diethyl TBS Gamma Cyclodexstrin based
Polarity Chiral stationary phase
Crossbond No
Temperature Limit 230 °C