Avantor® Hichrom HI-DEX DET Beta, GC Columns
Supplier: VWR International
HI68-25-015-25 HI68-25-015-30 HI68-25-015-50 HI68-25-015-60 HI68-25-025-15 HI68-25-025-25 HI68-25-025-30 HI68-25-025-50 HI68-25-025-60 HI80-10-010-10 HI80-10-010-5
76429-934EA 1461.26 USD
76429-934 76429-936 76429-938 76429-940 76429-942 76429-944 76429-946 76429-948 76429-950 76429-952 76430-070 76430-072
Avantor® Hichrom HI-DEX DET Beta, GC Columns
Chromatography Columns
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Order Now
SPECIFICATIONS
Phase name HI-DEX DET Beta
Environmentally Preferable
Phase composition Diethyl TBS Beta Cyclodexstrin based
Polarity Chiral stationary phase
Crossbond No
Temperature Limit 230 °C