Avantor® Hichrom HI-POF 2, GC Columns
Supplier: VWR International
76428-194 76428-196 76428-198 76428-200 76428-202 76428-204 76428-206 76428-208 76428-210 76428-212 76428-214
76428-194EA 1928.42 USD
76428-194 76428-196 76428-198 76428-200 76428-202 76428-204 76428-206 76428-208 76428-210 76428-212 76428-214
Avantor® Hichrom HI-POF 2, GC Columns
Chromatography Columns
Application specifc columns developed for pesticides, herbicides, insecticides analysis and so on.
Maximum temperature limits may change depending on the film thickness.
Order Now
SPECIFICATIONS
Environmentally Preferable
Phase name HI-POF 2
Phase composition Proprietary Phase developed for pesticides
Crossbond Yes