Avantor® Hichrom HI-ALC 1, GC Columns
Supplier: VWR International
76426-532 76426-534 76426-536
76426-532EA 1815.77 USD
76426-532 76426-534 76426-536
Avantor® Hichrom HI-ALC 1, GC Columns
Chromatography Columns
Application specific column for blood alcohols testing.
Max temperature limits may change depending on the film thickness.
Order Now
SPECIFICATIONS
Phase name HI-ALC 1
Environmentally Preferable
Phase composition Proprietary Phase developed for blood alcohol analysis
Crossbond Yes
Temperature Limit 280 °C