High-Performance Ovens, Forced Air, Sheldon Manufacturing
Supplier: Sheldon Manufacturing
SLFHP1022-H SLFHP1424-H SLFHP2822-H SLFHP3824-H SLFHP522-H
76423-292EA 12567.7 USD
76423-292 76423-294 76423-296 76423-298 76423-300
High-Performance Ovens, Forced Air, Sheldon Manufacturing
Ovens Standard Ovens
SHEL LAB High Performance Ovens are engineered to meet the most critical temperature requirements. They are designed for continuous drying operations at temperatures up to 260 or 306 °C depending on the model. These ovens have rugged, welded construction with a double-walled, corrosion-resistant stainless steel interior and a powder-coated exterior.
  • Horizontal air flow
  • Dual gasket door design
  • 40-step ramp and soak temperature controller or Watlow EZ-Zone heat up and soak controller for SMO14HP-2 and SMO38HP-2 models
  • Stainless steel interior
  • Built-in over
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Temperature Stability 0.3 °C