Corepoint Scientific™ Undercounter Controlled Room Temperature Cabinet, Freestanding with Solid Door, Horizon Scientific
Supplier: Horizon Scientific
CRTPR061WWW/0
76422-928EA 2229.77 USD
76422-928
Corepoint Scientific™ Undercounter Controlled Room Temperature Cabinet, Freestanding with Solid Door, Horizon Scientific
Testing Chambers Climatic Chambers
These CRT cabinets provide controlled room temperature environments for storing sensitive substances that require precise temperature management.
  • Integrated temperature alarms
  • Keyed door locks
  • Forced draft air circulation
  • Probe access
  • Precise microprocessor temperature control
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These specialized cabinets are ideal for scientific and medical applications. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency. Freestanding units require a 4-inch margin onMore Product Information