SteamScrubber® Glassware Washers, Labconco®
Supplier: Labconco
401001000 401001010 401001013 401101000 401101010 401101013 402001000 402001010 402001013 402101000 402101010 402101013
76420-684EA 12080 USD
76420-684 76420-686 76420-688 76420-690 76420-692 76420-694 76420-696 76420-698 76420-700 76420-702 76420-704 76420-706
SteamScrubber® Glassware Washers, Labconco®
Glassware Washers
Versatile and efficient glassware washers that can handle a wide range of wide-mouth beakers and general purpose labware.
  • Touch screen display with intuitive CleanWorks OS software
  • Flexible design allows for use of standard and injector spindle racks
  • Maximum wash temperature of 93 °C removes tough contaminants
  • Advanced wash pump maximizes water circulation up to 112 gal/min (424 liters/min)
  • Standard forced air chamber drying with SpeedVent technology
  • Durable Type 304 stainless steel chamber and racks
These are designed for efficient and consistentMore Product Information
Order Now
SPECIFICATIONS
Model SteamScrubber® free standing
Environmentally Preferable Waste Reducing,Water Efficient Waste Reducing,Water Efficient