Avantor® Seradigm, Select Grade Fetal Bovine Serum (FBS)
Supplier: VWR
1300-500 1300-500H 1300-050 1300-050H 1300-500CR 1300-050CR 1300-500CH 1300-050CH 1300-500CG 1300-500CGH 1300-050T 1300-050TH 1300-500T 1300-500TG 1300-500TH 1300-500G 1300-500GH
89510-186EA 497.47 USD
89510-186 89510-188 89510-182 89510-184 76419-584 76419-586 76419-588 76419-590 76509-316 76509-318 76509-320 76509-322 76509-324 76509-326 76509-328 77008-250 77008-252
Avantor® Seradigm, Select Grade Fetal Bovine Serum (FBS)
Sera (Serum)
Avantor Seradigm Select Grade FBS provides superior performance and consistency in cost-sensitive applications.
  • Industry-leading collection techniques at the source
  • Risk mitigation through a robust quality system
  • Proven track record for performance and consistency
  • Sourced from countries with health status classified as “Negligible BSE Risk” by the World Health Organization
The care and attention we devote to the collection and manufacture of our products generates quality that matters. Avantor Seradigm Select Grade FBS preserves more of theMore Product Information