UVP Crosslinker, CL-3000, Analytik Jena
Supplier: Analytik Jena US
849-95-0615-02 849-95-0615-04 849-95-0615-06 849-95-0615-01 849-95-0615-03 849-95-0615-05
76417-142EA 3133.59 USD
76417-142 76417-144 76417-146 76417-676 76417-678 76417-680
UVP Crosslinker, CL-3000, Analytik Jena
Lights UV Lights, Lamps and Bulbs
Ultraviolet light is considered an ancillary disinfectant to chemical disinfectants in the healthcare environment.
  • Perfect for delivering the recommended >1 J/cm² germicidal UV dose for surface disinfection
  • Crosslinking of DNA and RNA to nylon or nitrocellulose membranes
  • Microprocessor control provides precise UV dose control
  • Irradiation can be defined as energy (Joules/cm²) or time (seconds)
  • Bulbs: 8 W (6)
The high-precision CL-3000 crosslinker is capable of producing a cumulative dosage of 1 J/cm², this is considered well beyond the requirementMore Product Information