Digital Vortex Mixer, TX4, VELP Scientifica
Supplier: VELP SCIENTIFIC INC.
F202A0270
76416-204EA 613.99 USD
76416-204
Digital Vortex Mixer, TX4, VELP Scientifica
Shakers and Mixers Vortex Mixers
Digital vortex mixer with adjustable stirring speed, in continuous or with the exclusive IR sensor.
  • Digital control of vibration time and speed
  • Timed operation up to 999:59 minutes
  • Universal voltage/frequency model
  • Accessories fit by simple pressure
  • Optimum resistance to chemicals from technopolymer casing
  • Excellent stability with zinc alloy base
Velp
Patented infrared sensor technology ensures a drastic reduction in repetitive strain.
Certificaciones: IP Protection Class according to CEI EN 60259: IP 42 protection against particles and liquids.
More Product Information