SureSTART™ WebSeal™ Well Microtiter Plates, Round U, V and Flat-Bottom
Supplier: Thermo Scientific
60180-P330 60180-P332 60180-P333 60180-P334 60180-P336 60180-P338 60180-P340 60180-P341 60180-P342 60180-P343 60180-P345 60180-P350
76415-936PK 570.51 USD
76415-936 76415-938 76415-940 76415-942 76415-944 76415-946 76415-948 76415-950 76415-952 76415-954 76415-956 76415-958
SureSTART™ WebSeal™ Well Microtiter Plates, Round U, V and Flat-Bottom
Microplates Solid Supported Liquid/Liquid Extraction Plates (SLE)
SureSTART™ Level 3 WebSeal™ Plate 96-well round U, V, and flat-bottom microtiter plates are designed for regulated labs that require high performing products. Level 3 well plates are for high performance applications and are suitable for regulated labs using all solvent types including non-polar solvents.
  • Specification certified
  • Suitable for HPLC/UHPLC applications
  • Performance Level 3 products include unique WebSeal Plate+ glass-coated polypropylene plates and plates with glass inserts
  • Suitable for QA/QC applications using core detectors such as UV, DAD/
More Product Information