Laser Microdissection (LMD) System, Leica Microsystems
Supplier: Avantor
11600288 11505189 11505215 11600312 11505151
76414-892EA 211550.44 USD
76414-892 76414-894 76414-896 76414-898 76414-900 76463-316 76463-318 76463-320 76463-322 76463-324
Laser Microdissection (LMD) System, Leica Microsystems
Microtomes and Microtome Assemblies Microtomes
We move the laser, not the sample. And we use gravity for collection. LMD systems provides perfectly cut, contamination-free, analysis-ready dissectates.
  • Unique laser-guidance based dissection method to collect dissectates via gravity – contact-free and contamination-free
  • High-precision optics to steer the laser beam along the desired cut line, leaving clean cuts of contamination-free isolates
  • LMD SmartCut objectives made specifically for the LMD system
  • Save on consumables, as the Leica LMD utilizes standard PCR tubes to capture isolated samples. You will
More Product Information