Rockers 2D/3D, IKA Works
Supplier: IKA Works
0004002000 0004003000 0004000000 0004001000
76414-102EA 1511.76 USD
76414-102 76414-104 76414-106 76414-108
Rockers 2D/3D, IKA Works
Shakers and Mixers Rotating/Rocking Shakers
Rocking shaker with adjustable/fixed speed for smooth mixing tasks. Used for homogeneous mixing in flasks, culture flasks, Petri dishes and tubes. In addition these three-dimensional rockers with tumbling motion are ideal for the cultivation of cell cultures, DANN extractions, distributing of cells and staining and destaining of cells.
  • Different attachments enable use for a variety of applications
  • Usable at 4 to 50 °C in incubators
  • Easy operation with touch keypad
  • Suitable for continuous operation
  • Shaking diameter: 8° (angle - basic) and 0 to 15° (angle)
More Product Information