AlphaTec® EPDM Isolator Gloves, 85-501 Series, Ansell
Supplier: Ansell Healthcare
85501095 85501110
76408-488CS 9524.85 USD
76408-488 76408-490
AlphaTec® EPDM Isolator Gloves, 85-501 Series, Ansell
Gloves
Non-clean/non sterile EPDM RABS/Isolator gloves for use in 203 mm (8") ports.
  • Designed for repeated autoclave sterilization (up to 50 times)
  • Port size: 203 mm (8")
  • Comfortable and dexterous with great tactile sensitivity
  • Resistant against hydrogen peroxide solutions and common disinfecting chemicals
  • Resists aging from exposure to oxygen, UV rays, and ozone
  • Halogen-free: suitable for disposal by incineration
AlphaTec® EPDM RABS and Isolator gloves are designed from high quality material, in compliance with FDA CFR21. Comfortable and dexterous, More Product Information