Petrolatum Gauze Dressings, DUKAL™ Corporation
Supplier: Dukal
206 210 204-12 205-12 207-12 208-12 209-12
76407-506BX 76.52 USD
76407-506 76407-508 76407-890 76407-892 76407-894 76407-896 76407-898
Petrolatum Gauze Dressings, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Sterile dressings provide soothing non-adherent protection with more convenience and less mess.
  • Non-adherent petrolatum
  • Clings and conforms to all body contours
  • Non-toxic, non-irritating, guaranteed sterile
  • Not made with natural rubber latex
Dressings can be used as an initial layer in surgical dressings, on skin graft sites, treatment of first degree burns, abrasions, and as wound packing.
Order Now
SPECIFICATIONS
Environmentally Preferable