BioX Cryogenic Vials, Barcoded
Supplier: TWD Scientific
BX88-6051 BX88-6052 BX88-6053 BX88-6101 BX88-6102 BX88-6103 BX88-6151 BX88-6152 BX88-6153 BX88-9051 BX88-9052 BX88-9053 BX88-9151 BX88-9152 BX88-9153 BX88-6201 BX88-6202 BX88-6203 BX88-6502 BX88-6503
76404-402CS 340.57 USD
76404-402 76404-404 76404-406 76404-408 76404-410 76404-412 76404-414 76404-416 76404-418 76404-420 76404-422 76404-424 76404-426 76404-428 76404-430 76404-432 76404-434 76404-436 76404-440 76404-442
BioX Cryogenic Vials, Barcoded
Vials
Barcoded polypropylene cryogenic vials with colored caps.
  • Suitable for liquid nitrogen storage
  • Pre-barcoded
  • O-ring free
  • Self standing
  • White writing patch and graduations
Unique permanent barcodes for superior traceability. 1D and human-readable on the side are perfect for hand scanning and the 2D barcodes on the bottom make these vials automation friendly.
Precaución: Use in liquid phase LN₂ is not recommended.