Corepoint Scientific™ White Diamond Series Flammable Storage Freezers, Horizon Scientific
Supplier: Horizon Scientific
FF301WWW/0MTS
76402-962EA 12470.4 USD
76402-962
Corepoint Scientific™ White Diamond Series Flammable Storage Freezers, Horizon Scientific
Freezers Flammable Material Freezers
These White Diamond flammable storage freezers are designed to store and protect flammable substances in laboratory settings.
  • Premium touchscreen controller
  • Advanced data logging capabilities
  • Microprocessor temperature control
  • Integrated temperature alarms
  • Digital 24-hour temperature chart
  • Keyed door lock
  • Natural hydrocarbon refrigerants
  • Manual defrosting
  • Meet NFPA guidelines
This line showcases elite features such as state-of-the-art touchscreen controllers, advanced alarms, data logging capabilities and unparalleled temperature control. All unitsMore Product Information