SP Bel-Art® Sterileware® Powder Thief, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37874-0001 F37874-0005 F37874-0010 F37874-0020 F37874-1010 F37874-1020
76402-906PK 261.88 USD
76402-906 76402-908 76402-910 76402-912 76402-914 76402-916 76402-918 76402-920
SP Bel-Art® Sterileware® Powder Thief, Bel-Art Products, a part of SP
Samplers Solid Samplers
These sterile powder thieves easily sample from targeted locations within bulk powders. Used by pharmaceutical, food, cosmetic and chemical companies.
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10⁻⁶)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Available in two lengths each with four tip volumes.
Información paraMore Product Information