SP Bel-Art® Sterileware® Liquid Thief, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37871-0500 F37871-1000
76402-902PK 328.37 USD
76402-902 76402-904
SP Bel-Art® Sterileware® Liquid Thief, Bel-Art Products, a part of SP
Samplers Fluid Samplers
These sterile liquid thieves draw a sample with a simple pull of the handle, then dispense easily into the vessel of your choice.
  • Ideal for sampling water and other low viscosity liquids, oils and ointments; 0.15 cm (¹⁄₁₆") tip hole diameter
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10⁻⁶)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for
More Product Information