Laboratory Glassware Washers, Model PLW 8617, Miele
Supplier: Miele
11179780 11179790 11179800 11179810 11179820 11179830 11179840 11179850
76402-688EA 61894.73 USD
76402-688 76402-690 76402-692 76402-694 76402-696 76402-698 76402-700 76402-702
Laboratory Glassware Washers, Model PLW 8617, Miele
Glassware Washers
The PLW 8617 laboratory washer can accommodate 216 laboratory flasks, 588 vials, or 294 pipettes. The large glassware washer is flexible and simple to use, it has modular load carriers and SimpleLoad system. This efficient unit has a single door, 1150 mm (3' 91/4") wide, a usable capacity of 351 litres. PLW 8617 provides reliable results, high pump performance with a variable-speed pump and features spray arm monitoring and a conductivity meter.
  • Innovative 2-pump system for maximum performance
  • New flexible and modular (SimpleLoad)racking system
  • Excellent
More Product Information