Polygosil C18 60-5, HPLC Columns
Supplier: Avantor
POLY-5C18-150A
76402-300EA 873.19 USD
76402-300 76402-302 76402-314
Polygosil C18 60-5, HPLC Columns
Chromatography Columns
Irregular silica phases for analytical applications.
  • pH stability: 2 to 8
Column hardware: Stainless steel.