BRAND Transferpette® S, Single Channel Pipettes, Mechanical, BrandTech
Supplier: Brandtech
705808 705816 705820 705862 705854 705864 705844 705838 705828 705868 705884 705882 705880 705878 705874 705873 705872 705870 705869 705890 705897 705896 705893 705892 705891
76383-050EA 269.96 USD
76383-050 76383-052 76383-054 76383-056 76383-058 76383-060 76383-062 76383-064 76383-066 76383-068 76383-070 76383-072 76383-074 76383-076 76383-078 76383-080 76383-082 76383-084 76383-086 76383-088 76383-090 76383-092 76383-094 76383-096 76383-098
BRAND Transferpette® S, Single Channel Pipettes, Mechanical, BrandTech
Pipettes Micropipettes
The Transferpette® S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Due to its perfect ergonomics, the Transferpette® S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands.
  • True one-hand operation for right- and left-handed users: Easily set the volume, pipette and eject the tip
  • Colour code on volume adjustment wheel, shaft and volume-change protection
  • Low operating forces and a short stroke ensure relaxed pipetting, even during
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Sustainable Packaging,Low Manufacturing Impact Sustainable Packaging