Avantor® ACE® C18-AR, HPLC Columns, 5 µm
Supplier: Avantor
ACE-129-2546
76382-586EA 1247.07 USD
76382-586
Avantor® ACE® C18-AR, HPLC Columns, 5 µm
Chromatography Columns
The Avantor® ACE® C18-AR phase utilizes a specially developed ligand combining a C18 chain with integral phenyl functionality, thus combining the benefits of both C18 and phenyl characteristics into a single phase. This can be used for ‘standard’ C18 column separations but is additionally recommended for separations that involve compounds containing aromatic functionality.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 10 µm) and
More Product Information