Orum International TRIO.BAS™ AIRBIO DUO Cable, Contact Plate, Hardy Diagnostics
Supplier: Hardy Diagnostics
BAS479K BAS481K
76377-536EA 16034.08 USD
76377-536 76377-540
Orum International TRIO.BAS™ AIRBIO DUO Cable, Contact Plate, Hardy Diagnostics
Samplers Air Samplers
The TRIO.BAS™ instruments utilize the impaction method of air sampling. The air, passing through an aspirating head, impacts onto an appropriate agar plate that collects bacteria or other organic substances possibly present in the sampled area.
  • Designed for benchtop/cart stability and oriented in a vertical position to capture laminar flow from the ceiling
  • Use separate culture media at the same time to determine bacterial and fungal counts
  • 100 or 200 liter per minute formats, contact plate or Petri plate formats
  • Charges via cable; Sample up to 70,000
More Product Information