Corepoint Scientific™ White Diamond Series Undercounter Built-In Refrigerator, Horizon Scientific
Supplier: Horizon Scientific
PR031WWW/0FB PR031WWW/0FBLH
76355-492EA 2169.56 USD
76355-492 76355-494
Corepoint Scientific™ White Diamond Series Undercounter Built-In Refrigerator, Horizon Scientific
Refrigerators
These undercounter laboratory refrigerators provide premium laboratory cold storage for scientific and medical use.
  • ntegrated temperature alarms
  • Keyed door locks
  • Forced draft circulation
  • Precise microprocessor temperature control
  • Natural hydrocarbon refrigerants
Built-in units are designed with front-facing ventilation systems, allowing easy installation into a bench or countertop. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Información para pedidos: All units have a three-year parts and laborMore Product Information