CORIO™ CP Heating Circulator Baths, JULABO USA, Inc.
Supplier: JULABO
9013504.22 9013504.33 9013506.22 9013506.33 9013512.22 9013512.33 9013526.22 9013526.33
76353-820EA 5244.24 USD
76353-820 76353-822 76353-824 76353-826 76353-828 76353-830 76353-832 76353-834
CORIO™ CP Heating Circulator Baths, JULABO USA, Inc.
Circulating Thermostats Open Circulator Systems Baths with Integral Circulators
CORIO™ CP heating circulator combinations with stainless steel bath tanks for internal or external temperature applications from 20 to 200 °C.
  • Infinitely adjustable pressure pump
  • Easy to switch between internal and external circulation
  • Bright LED display
  • USB and RS232 communication ports
For internal and external temperature applications. High quality stainless steel bath tanks with bath lid and drain tap.
CORIO CP circulators feature bright LED display with push button control. A lever below the display enables simple adjustment between internalMore Product Information
Order Now
SPECIFICATIONS
Model CP-BC4 CP-BC6 CP-BC12 CP-BC26
Environmentally Preferable
Pump Flow Rate 8 - 27 L/min 8 - 27 L/min 8 - 27 L/min 8 - 27 L/min
Pump Pressure 1.5 - 10.2 psi 1.5 - 10.2 psi 1.5 - 10.2 psi 1.5 - 10.2 psi