PHCbi MPR Series Pharmaceutical Refrigerators with Freezer, PHC Corporation
Supplier: PHC Corporation
MPR-N450FH-PA MPR-N450FSH-PA MPR-N250FH-PA MPR-N250FSH-PA
76353-814EA 10003.7 USD
76353-814 76353-816 76498-952 76498-954
PHCbi MPR Series Pharmaceutical Refrigerators with Freezer, PHC Corporation
Refrigerators
The PHCbi brand Pharmaceutical Refrigerators with Freezer offers a complete storage solution in one cabinet. These multipurpose cabinets are designed for safe, reliable storage of vaccines and biological materials.
  • Refrigerated and frozen products can be maintained within a single footprint
  • Optimum temperature uniformity under full load conditions
  • Independent refrigeration systems
  • The microprocessor controller and softkey interface are door mounted at eye level
  • Adjustable audible and visual alarms
  • Slim profile cabinet design
  • Data transfer is through a USB
More Product Information