D-(+)-Maltose monohydrate ≥85%

Supplier: RPI

Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate

M22000-100.0 M22000-1000.0 M22000-20000.0 M22000-250.0 M22000-500.0 M22000-5000.0
76346-778EA 39.4 USD
76346-778 76346-780 76346-782 76346-784 76346-786 76346-788
D-(+)-Maltose monohydrate ≥85%
D-(+)-Maltose monohydrate

Precaución: Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₁₂H₂₂O₁₁·1H₂O
MW: 360.32 g/mol
Melting Pt: 119…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00149343
CAS Number: 6363-53-7
EINECS: 200-716-5
Merck Index: 13,05736
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f861d7adf53134a0c485b5942d3be34c4d0b571&vid=16e5c4425a7a8e158a4200470a45483c7ff5781e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance: White powder, odorless and no unusual taste
Water: ≤ 7 %
Purity: ≥ 85 %
Particle Size: ≤ 1.70 mm
Process Of Manufacture Extraction
Total Aerobic Microbial Count: ≤ 300 CFU/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR