5-Bromo-2'-deoxyuridine (broxuridine)

Supplier: RPI
Warning

Synonyms: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU

B74200-0.25 B74200-0.5 B74200-1.0 B74200-5.0
76345-300EA 28.61 USD
76345-300 76345-302 76345-304 76345-306
5-Bromo-2'-deoxyuridine (broxuridine)
5-Bromo-2'-deoxyuridine (broxuridine)

Thymidine analog used for study of DNA synthesis.


This analog selectively incorporates into cellular DNA during S-phase. Supplied as white powder.

Formula: C₉H₁₁BrN₂O₅
MW: 307.10 g/mol
Melting Pt: 191…194 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006529
CAS Number: 59-14-3
EINECS: 200-415-9

Order Now

Specification Test Results

Appearance White powder
Purity (HPLC) ≥ 99 %
Water Content (K.F) ≤ 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR